Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-2-fluorophenol ≥95% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

001746-5G 001746-25G
101782-462EA 23.08 USD InStock
101782-462 101782-464
4-Amino-2-fluorophenol ≥95% for synthesis
4-Amino-2-fluorophenol
Hazard code(s): Irritant, Refrigerate


Formula: C₆H₆FNO
MW: 127.12
Melting Pt: 170…172 °C
MDL Number: MFCD00671760
CAS Number: 399-96-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty