Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-4-methoxybenzoic acid ≥98%

Supplier: MATRIX SCIENTIFIC MS

001793-5G 001793-25G
101782-590EA 29.23 USD InStock
101782-590 102550-802
3-Bromo-4-methoxybenzoic acid ≥98%
3-Bromo-4-methoxybenzoic acid
Hazard code(s): Irritant


Formula: C₈H₇BrO₃
Melting Pt: 216…217 °C
MDL Number: MFCD00020295
CAS Number: 99-58-1
EINECS: 202-768-4
Order Now
Specification Test Results
Melting point 216-217°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty