Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethyl-3-methoxypyrazine ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

001801-1G 001801-5G 001801-25G
101782-624EA 27.69 USD InStock
101782-624 101782-626 101782-628
2-Ethyl-3-methoxypyrazine ≥98% for synthesis
2-Ethyl-3-methoxypyrazine
Hazard code(s): Irritant


Formula: C₇H₁₀N₂O
MW: 138.17
Boiling Pt: 187 °C
Density: 1.029 g/cm³
MDL Number: MFCD00006129
CAS Number: 25680-58-4
EINECS: 247-184-0
Order Now
Specification Test Results
Boiling point 187°C
Density 1.029
ND20 1.502

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty