Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Iodoperfluorohexane ≥99% stabilised for synthesis

Supplier: MATRIX SCIENTIFIC MS

001838-25G 001838-100G
101782-694EA 44.62 USD InStock
101782-694 102832-740
1-Iodoperfluorohexane ≥99% stabilised for synthesis
1-Iodoperfluorohexane
Hazard code(s): Irritant


Formula: C₆F₁₃I
MW: 445.95 g/mol
Boiling Pt: 117
Melting Pt: -45
Density: 2.063 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00001063
CAS Number: 355-43-1
EINECS: 206-586-6
Order Now
Specification Test Results
Boiling point 115-117°C
Density 2.063
ND20 1.327

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty