Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2-fluorophenol ≥97%

Supplier: MATRIX SCIENTIFIC MS

002111-5G 002111-25G 002111-100G
101783-220EA 18.46 USD InStock
101783-220 101783-222 101783-224
3-Chloro-2-fluorophenol ≥97%
3-Chloro-2-fluorophenol
Hazard code(s): Irritant, Irritant-Harmful


Formula: C₆H₄ClFO
MDL Number: MFCD01631572
CAS Number: 2613-22-1
UN: 3261
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty