Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluorosalicylic acid ≥97% for synthesis

Supplier: MATRIX SCIENTIFIC MS

002123-5G 002123-25G
101783-250EA 44.62 USD InStock
101783-250 101783-252
5-Fluorosalicylic acid ≥97% for synthesis
5-Fluorosalicylic acid
Hazard code(s): Irritant


Formula: C₇H₅FO₃
MW: 156.11 g/mol
Boiling Pt: 295
Melting Pt: 180
MDL Number: MFCD00002456
CAS Number: 345-16-4
EINECS: 206-455-3
Order Now
Specification Test Results
Melting point 177-179°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty