Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-o-cresol ≥98%

Supplier: MATRIX SCIENTIFIC MS
Warning

002184-5G 002184-25G
101783-418EA 112.31 USD InStock
101783-418 101783-420
5-Chloro-o-cresol ≥98%
5-Chloro-o-cresol
Hazard code(s): Irritant


Formula: C₇H₇ClO
Boiling Pt: 225 °C (1013 hPa)
Melting Pt: 75 - 76 °C
MDL Number: MFCD00060672
CAS Number: 5306-98-9
EINECS: 226-160-3
UN: 3437
ADR: 6.1,II
Order Now
Specification Test Results
Melting point 73°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty