Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H,1H,2H,2H-Tridecafluoro-1-octanol ≥97%

Supplier: MATRIX SCIENTIFIC MS

002240-25G 002240-100G
101783-506EA 64.62 USD InStock
101783-506 101783-508
1H,1H,2H,2H-Tridecafluoro-1-octanol ≥97%
1H,1H,2H,2H-Tridecafluoro-1-octanol
Hazard code(s): Irritant


Formula: C₈H₅F₁₃O
Boiling Pt: 91…93 °C (36mm)
Density: 1.680 g/cm³
MDL Number: MFCD00042143
CAS Number: 647-42-7
EINECS: 211-477-1
Order Now
Specification Test Results
Boiling point 91-93°C/36m
Density 1.68
ND20 1.314

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty