Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α,α,α,2-Tetrafluoro-m-toluic acid ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

002281-5G 002281-25G
101783-598EA 26.15 USD InStock
101783-598 101783-600
α,α,α,2-Tetrafluoro-m-toluic acid ≥98% for synthesis
α,α,α,2-Tetrafluoro-m-toluic acid
Hazard code(s): Irritant


Formula: C₈H₄F₄O₂
Melting Pt: 126…128 °C
MDL Number: MFCD00040980
CAS Number: 115029-22-6
Order Now
Specification Test Results
Melting point 126-128°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty