Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-3-(trifluoromethyl)benzoic acid ≥97% for synthesis

Supplier: MATRIX SCIENTIFIC MS

002297-1G 002297-5G
101783-634EA 20 USD InStock
101783-634 101783-636
2-Chloro-3-(trifluoromethyl)benzoic acid ≥97% for synthesis
2-Chloro-3-(trifluoromethyl)benzoic acid
Hazard code(s): Irritant


Formula: C₈H₄ClF₃O₂
MW: 224.57
MDL Number: MFCD00792415
CAS Number: 39226-97-6
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty