Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-3-nitrobenzoic acid ≥97% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

002393-25G 002393-100G
101783-824EA 29.23 USD InStock
101783-824 101783-826
4-Fluoro-3-nitrobenzoic acid ≥97% for synthesis
4-Fluoro-3-nitrobenzoic acid
Hazard code(s): Irritant


Formula: C₇H₄FNO₄
MW: 185.11 g/mol
Boiling Pt: 92
Melting Pt: 119.5…121 °C
MDL Number: MFCD00007058
CAS Number: 453-71-4
EINECS: 207-221-3
Order Now
Specification Test Results
Boiling point 92°C/15mm
Melting point 119.5-121°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty