Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4,5-difluorobenzoic acid ≥99%

Supplier: MATRIX SCIENTIFIC MS
Danger

002414-5G 002414-25G 002414-100G
101783-862EA 26.15 USD InStock
101783-862 101783-864 101783-866
2-Chloro-4,5-difluorobenzoic acid ≥99%
2-Chloro-4,5-difluorobenzoic acid
Hazard code(s): Irritant


Formula: C₇H₃ClF₂O₂
Boiling Pt: 258 °C (1013 hPa)
Melting Pt: 103 - 106 °C
Storage Temperature: Ambient
MDL Number: MFCD00077479
CAS Number: 110877-64-0
EINECS: 405-380-5
Order Now
Specification Test Results
Melting point 103-106°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty