Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4,5-dimethoxybenzoic acid ≥98%

Supplier: MATRIX SCIENTIFIC MS

002418-1G 002418-5G
101783-876EA 43.08 USD InStock
101783-876 101783-878
2-Chloro-4,5-dimethoxybenzoic acid ≥98%
2-Chloro-4,5-dimethoxybenzoic acid
Hazard code(s): Irritant


Formula: C₉H₉ClO₄
Melting Pt: 183…185 °C
MDL Number: MFCD00079727
CAS Number: 60032-95-3
Order Now
Specification Test Results
Melting point 183-185°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty