Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Pyridineboronic acid ≥95% for synthesis

Supplier: MATRIX SCIENTIFIC MS

002475-25G 002475-100G
101783-952EA 78.46 USD InStock
101783-952 101783-954
4-Pyridineboronic acid ≥95% for synthesis
4-Pyridineboronic acid
Hazard code(s): Irritant, Store in Freezer


Formula: C₅H₆BNO₂
MDL Number: MFCD01074545
CAS Number: 1692-15-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty