Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2,4-dimethoxybenzoate ≥98%

Supplier: MATRIX SCIENTIFIC MS

002485-5G 002485-25G
101783-968EA 66.15 USD InStock
101783-968 101783-970
Methyl-2,4-dimethoxybenzoate ≥98%
Methyl-2,4-dimethoxybenzoate
Hazard code(s): Irritant


Formula: C₁₀H₁₂O₄
Boiling Pt: 110 °C (1mm)
MDL Number: MFCD00070649
CAS Number: 2150-41-6
Order Now
Specification Test Results
Boiling point 110°C/1mm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty