Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-fluorobenzamide ≥97%

Supplier: MATRIX SCIENTIFIC MS

002494-1G 002494-5G
101783-990EA 38.46 USD InStock
101783-990 101783-992
2-Chloro-4-fluorobenzamide ≥97%
2-Chloro-4-fluorobenzamide
Hazard code(s): Irritant


Formula: C₇H₅ClFNO
Melting Pt: 147…149 °C
MDL Number: MFCD00042531
CAS Number: 88578-90-9
Order Now
Specification Test Results
Melting point 147-149°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty