Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-iodoaniline ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

002544-25G 002544-100G
101784-124EA 29.23 USD InStock
101784-124 101784-126
2-Chloro-4-iodoaniline ≥98% for synthesis
2-Chloro-4-iodoaniline
Hazard code(s): Irritant


Formula: C₆H₅ClIN
MW: 253.47
Boiling Pt: 289 °C (1013 hPa)
Melting Pt: 68…70 °C
MDL Number: MFCD00025298
CAS Number: 42016-93-3
UN: 2811
ADR: 6.1 III
Order Now
Specification Test Results
Melting point 68-70°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty