Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-p-anisaldehyde ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

002554-5G 002554-25G 002554-100G
101784-154EA 32.31 USD InStock
101784-154 101784-156 101784-158
2-Fluoro-p-anisaldehyde ≥98% for synthesis
2-Fluoro-p-anisaldehyde
Hazard code(s): Irritant, Air Sensitive


Formula: C₈H₇FO₂
MW: 154.14 g/mol
Melting Pt: 44…46 °C
MDL Number: MFCD00236679
CAS Number: 331-64-6
Order Now
Specification Test Results
Melting point 44-46°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty