Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-nitroanisole ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

002590-5G 002590-25G
101784-228EA 13.85 USD InStock
101784-228 101784-230
2-Chloro-4-nitroanisole ≥98% for synthesis
2-Chloro-4-nitroanisole
Hazard code(s): Irritant


Formula: C₇H₆ClNO₃
Melting Pt: 94…96 °C
MDL Number: MFCD00060681
CAS Number: 4920-79-0
EINECS: 225-547-4
Order Now
Specification Test Results
Melting point 94-96°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty