Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-5-(trifluoromethyl)benzyl alcohol ≥97% for synthesis

Supplier: MATRIX SCIENTIFIC MS

002629-1G 002629-5G
101784-344EA 30.77 USD InStock
101784-344 101784-346
3-Fluoro-5-(trifluoromethyl)benzyl alcohol ≥97% for synthesis
3-Fluoro-5-(trifluoromethyl)benzyl alcohol
Hazard code(s): Irritant


Formula: C₈H₆F₄O
MW: 194.13
MDL Number: MFCD00061169
CAS Number: 184970-29-4
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty