Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-(trifluoromethyl)phenyl isothiocyanate ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

002672-5G 002672-25G
101784-404EA 95.38 USD InStock
101784-404 101784-406
2-Chloro-5-(trifluoromethyl)phenyl isothiocyanate ≥98% for synthesis
2-Chloro-5-(trifluoromethyl)phenyl isothiocyanate
Hazard code(s): Toxic, Corrosive, Lachrymator, Refrigerate, Moisture Sensitive


Formula: C₈H₃ClF₃NS
MW: 237.63 g/mol
Boiling Pt: 126…127 °C (35mm)
Density: 1.450 g/cm³
MDL Number: MFCD00041055
CAS Number: 23165-49-3
Order Now
Specification Test Results
Boiling point 126-127°C/3
Density 1.45

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty