Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-fluoroanisole for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

002764-1G 002764-5G
101784-610EA 13.85 USD InStock
101784-610 101784-612
2-Chloro-6-fluoroanisole for synthesis
2-Chloro-6-fluoroanisole


Formula: C₇H₆ClFO
MDL Number: MFCD01631573
CAS Number: 53145-38-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty