Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Fluoropicolinic acid ≥97%

Supplier: MATRIX SCIENTIFIC MS

002799-5G 002799-25G
101784-692EA 60 USD InStock
101784-692 101784-694
6-Fluoropicolinic acid ≥97%
6-Fluoropicolinic acid
Hazard code(s): Irritant


Formula: C₆H₄FNO₂
Melting Pt: 135…137 °C
MDL Number: MFCD02181193
CAS Number: 402-69-7
Order Now
Specification Test Results
Melting point 135-137°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty