Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H,1H,7H-Dodecafluoro-1-heptanol ≥97%

Supplier: MATRIX SCIENTIFIC MS

002822-25G 002822-100G
101784-722EA 36.92 USD InStock
101784-722 101784-724
1H,1H,7H-Dodecafluoro-1-heptanol ≥97%
1H,1H,7H-Dodecafluoro-1-heptanol
MDL: MFCD00039630


Formula: C₇H₄F₁₂O
MDL Number: MFCD00039630
CAS Number: 335-99-9
EINECS: 206-406-6
Order Now
Specification Test Results
Boiling point 169-170°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty