Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium trifluoroacetate ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

002907-25G 002907-100G
101784-892EA 16.92 USD InStock
101784-892 101784-894
Potassium trifluoroacetate ≥98% for synthesis
Potassium trifluoroacetate
Hazard code(s): Marine Pollutant, Irritant-Harmful, Hygroscopic


Formula: C₂F₃KO₂
MW: 152.11 g/mol
Melting Pt: 143…145 °C
MDL Number: MFCD00013215
CAS Number: 2923-16-2
EINECS: 220-877-5
Order Now
Specification Test Results
Melting point 143-145°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty