Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2'-Fluoroacetophenone ≥97% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

002921-25G 002921-100G
101784-914EA 13.85 USD InStock
101784-914 101784-916
2'-Fluoroacetophenone ≥97% for synthesis
2'-Fluoroacetophenone
Hazard code(s): Irritant


Formula: C₈H₇FO
MW: 138.14 g/mol
Boiling Pt: 63 °C (9mm)
Density: 1.121 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000320
CAS Number: 445-27-2
EINECS: 207-156-0
Order Now
Specification Test Results
Boiling point 63°C/9mm
Density 1.121
ND20 1.5075

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty