Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Acetoxybenzaldehyde ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

003000-5G 003000-25G
101785-038EA 120 USD InStock
101785-038 101785-040
3-Acetoxybenzaldehyde ≥98% for synthesis
3-Acetoxybenzaldehyde
Hazard code(s): Irritant


Formula: C₉H₈O₃
Boiling Pt: 138 °C (20mm)
MDL Number: MFCD00016603
CAS Number: 34231-78-2
EINECS: 251-890-4
Order Now
Specification Test Results
Boiling point 138/20mm

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty