Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Acetoxybenzaldehyde ≥95% for synthesis

Supplier: MATRIX SCIENTIFIC MS

003002-25G 003002-100G 003002-500G
101785-042EA 24.62 USD InStock
101785-042 101785-044 101785-046
4-Acetoxybenzaldehyde ≥95% for synthesis
4-Acetoxybenzaldehyde
Hazard code(s): Irritant


Formula: C₉H₈O₃
Boiling Pt: 152…153 °C (17mm)
Density: 1.168 g/cm³
MDL Number: MFCD00003384
CAS Number: 878-00-2
EINECS: 212-898-3
Order Now
Specification Test Results
Boiling point 152-153°C/1
Density 1.168
ND20 1.538

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty