Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chloroacetamidine hydrochloride ≥97% for synthesis

Supplier: MATRIX SCIENTIFIC MS

003066-1G 003066-5G 003066-25G
101785-148EA 29.23 USD InStock
101785-148 101785-150 101785-152
Chloroacetamidine hydrochloride ≥97% for synthesis
Chloroacetamidine hydrochloride
Hazard code(s): Irritant, Moisture Sensitive


Formula: C₂H₆Cl₂N₂
Melting Pt: 98…99 °C
MDL Number: MFCD00053013
CAS Number: 10300-69-3
EINECS: 233-675-7
UN: 3261
ADR: 8,III
Order Now
Specification Test Results
Melting point 98-99°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty