Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3'-Aminoacetophenone ≥97% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

003071-100G 003071-500G
101785-158EA 73.85 USD InStock
101785-158 101785-160
3'-Aminoacetophenone ≥97% for synthesis
3'-Aminoacetophenone
Hazard code(s): Irritant


Formula: C₈H₉NO
MW: 135.17 g/mol
Boiling Pt: 289…290 °C
Melting Pt: 96…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00007796
CAS Number: 99-03-6
EINECS: 202-722-3
Merck Index: 13,00412
Order Now
Specification Test Results
Boiling point 289-290°C
Melting point 96-98°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty