Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Cyanophenyl isothiocyanate ≥98%

Supplier: MATRIX SCIENTIFIC MS

003208-1G 003208-5G
101785-408EA 36.92 USD InStock
101785-408 101785-410
2-Cyanophenyl isothiocyanate ≥98%
2-Cyanophenyl isothiocyanate
Hazard code(s): Irritant, Lachrymator, Refrigerate


Formula: C₈H₄N₂S
Melting Pt: 60…61 °C
MDL Number: MFCD00060361
CAS Number: 81431-98-3
Order Now
Specification Test Results
Melting point 60-61°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty