Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Nitrophenyl)acetonitrile ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

003238-1G 003238-5G
101785-432EA 23.08 USD InStock
101785-432 101785-434
(3-Nitrophenyl)acetonitrile ≥98% for synthesis
(3-Nitrophenyl)acetonitrile
Hazard code(s): Irritant-Harmful


Formula: C₈H₆N₂O₂
Boiling Pt: 145 - 150 °C (0.5 -rr)
Melting Pt: 63 °C
MDL Number: MFCD00041249
CAS Number: 621-50-1
EINECS: 210-689-1
Order Now
Specification Test Results
Melting point 63°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty