Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Ethoxybenzaldehyde ≥99%

Supplier: MATRIX SCIENTIFIC MS

003258-5G 003258-25G
101785-462EA 73.85 USD InStock
101785-462 101785-464
3-Ethoxybenzaldehyde ≥99%
3-Ethoxybenzaldehyde
Hazard code(s): Irritant, Air Sensitive


Formula: C₉H₁₀O₂
Boiling Pt: 243 °C (1013 hPa)
Melting Pt: 21 °C
Density: 1.06 g/cm³ (20 °C)
MDL Number: MFCD00016606
CAS Number: 22924-15-8
EINECS: 245-333-4
Order Now
Specification Test Results
Boiling point 245°C
Density 1.08

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty