Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluorobenzoic acid ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

003292-100G 003292-500G
101785-530EA 60 USD InStock
101785-530 101785-532
4-Fluorobenzoic acid ≥98% for synthesis
4-Fluorobenzoic acid
Hazard code(s): Irritant


Formula: C₇H₅FO₂
Boiling Pt: 254 °C (1013 hPa)
Melting Pt: 185 °C
Density: 1.49 g/cm³ (20 °C)
Flash Pt: ≥ 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002530
CAS Number: 456-22-4
EINECS: 207-259-0
Merck Index: 13,04198
Order Now
Specification Test Results
Melting point 182-184°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty