Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluorobenzophenone ≥98%

Supplier: MATRIX SCIENTIFIC MS
Warning

003301-25G 003301-100G
101785-558EA 23.08 USD InStock
101785-558 101785-560
2-Fluorobenzophenone ≥98%
2-Fluorobenzophenone
Hazard code(s): Irritant


Formula: C₁₃H₉FO
MW: 200.21 g/mol
Boiling Pt: 190 °C (29mm)
Density: 1.186 g/cm³
MDL Number: MFCD00000318
CAS Number: 342-24-5
EINECS: 206-440-1
Order Now
Specification Test Results
Boiling point 190°C/29mm
Density 1.186
ND20 1.5864

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty