Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrabromothiophene ≥98%

Supplier: MATRIX SCIENTIFIC MS

003430-25G 003430-100G
101785-704EA 52.31 USD InStock
101785-704 101785-706
Tetrabromothiophene ≥98%
Tetrabromothiophene
Hazard code(s): Irritant


Formula: C₄Br₄S
Boiling Pt: 326 °C
Melting Pt: 116…118 °C
MDL Number: MFCD00005419
CAS Number: 3958-03-0
EINECS: 223-554-7
Order Now
Specification Test Results
Boiling point 326°C
Melting point 116-118°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty