Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3,5,6-Tetrafluoropyridine ≥98%

Supplier: MATRIX SCIENTIFIC MS
Danger

003463-1G 003463-5G 003463-25G
101785-736EA 26.15 USD InStock
101785-736 101785-738 102550-828
2,3,5,6-Tetrafluoropyridine ≥98%
2,3,5,6-Tetrafluoropyridine
Hazard code(s): Flammable


Formula: C₅HF₄N
Boiling Pt: 102…103 °C
Density: 1.499 g/cm³ (20 °C)
MDL Number: MFCD00011734
CAS Number: 2875-18-5
UN: 1993
ADR: 3,III
Order Now
Specification Test Results
Boiling point 102-103°C
Density 1.499
ND20 1.4046

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty