Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H,1H-Tridecafluoro-1-heptanol ≥97% for synthesis

Supplier: MATRIX SCIENTIFIC MS

003557-5G 003557-25G 003557-100G
101785-870EA 50.77 USD InStock
101785-870 101785-872 101785-874
1H,1H-Tridecafluoro-1-heptanol ≥97% for synthesis
1H,1H-Tridecafluoro-1-heptanol
Hazard code(s): Irritant


Formula: C₇H₃F₁₃O
MW: 350.08 g/mol
Boiling Pt: 147 °C
MDL Number: MFCD00042433
CAS Number: 375-82-6
EINECS: 206-796-8
Order Now
Specification Test Results
Boiling point 147°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty