Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4,5-Trifluorobenzoic acid ≥98%

Supplier: MATRIX SCIENTIFIC MS
Warning

003630-5G 003630-25G
101785-964EA 16.92 USD InStock
101785-964 101785-966
3,4,5-Trifluorobenzoic acid ≥98%
3,4,5-Trifluorobenzoic acid
Hazard code(s): Irritant


Formula: C₇H₃F₃O₂
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 97…99 °C
MDL Number: MFCD00074962
CAS Number: 121602-93-5
Order Now
Specification Test Results
Melting point 97-99°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty