Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Acetylphenylboronic acid ≥97% for synthesis

Supplier: MATRIX SCIENTIFIC MS

003910-5G 003910-25G
101786-424EA 36.92 USD InStock
101786-424 101786-426
3-Acetylphenylboronic acid ≥97% for synthesis
3-Acetylphenylboronic acid
Hazard code(s): Irritant, Store in Freezer


Formula: C₈H₉BO₃
Melting Pt: 204…206 °C
MDL Number: MFCD01074678
CAS Number: 204841-19-0
Order Now
Specification Test Results
Melting point 204-206°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty