Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl hexafluoroglutarate ≥98%

Supplier: MATRIX SCIENTIFIC MS

003940-25G 003940-100G
101786-482EA 129.23 USD InStock
101786-482 101786-484
Dimethyl hexafluoroglutarate ≥98%
Dimethyl hexafluoroglutarate
Hazard code(s): Irritant


Formula: C₇H₆F₆O₄
MDL Number: MFCD00155851
CAS Number: 1513-62-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty