Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chlorothioanisole ≥99% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

003953-5G 003953-25G 003953-100G
101786-514EA 26.15 USD InStock
101786-514 101786-516 101786-518
2-Chlorothioanisole ≥99% for synthesis
2-Chlorothioanisole
Hazard code(s): Stench


Formula: C₇H₇ClS
MW: 158.65
Boiling Pt: 116…117 °C (15mm)
MDL Number: MFCD00040124
CAS Number: 17733-22-1
UN: 3334
ADR: 9
Order Now
Specification Test Results
Boiling point 116-117°C/1

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty