Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-thiopheneboronic acid ≥97% for synthesis

Supplier: MATRIX SCIENTIFIC MS

003966-1G 003966-5G
101786-532EA 16.92 USD InStock
101786-532 101786-534
5-Chloro-2-thiopheneboronic acid ≥97% for synthesis
5-Chloro-2-thiopheneboronic acid
Hazard code(s): Irritant, Refrigerate


Formula: C₄H₄BClO₂S
MW: 162.4
MDL Number: MFCD00236030
CAS Number: 162607-18-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty