Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2,5-Dimethoxyphenyl)acetic acid ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

004016-5G 004016-25G
101786-668EA 121.54 USD InStock
101786-668 101786-670
(2,5-Dimethoxyphenyl)acetic acid ≥98% for synthesis
(2,5-Dimethoxyphenyl)acetic acid
Hazard code(s): Irritant


Formula: (C₆H₃(OCH₃)₂)CH₂COOH
MW: 196.2 g/mol
Boiling Pt: 327 °C (1013 hPa)
Melting Pt: 123…125 °C
MDL Number: MFCD00004322
CAS Number: 1758-25-4
EINECS: 217-151-5
Order Now
Specification Test Results
Melting point 123-125°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty