Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2,3-Dimethoxyphenyl)acetonitrile ≥98%

Supplier: MATRIX SCIENTIFIC MS

004022-1G 004022-5G
101786-690EA 52.31 USD InStock
101786-690 101786-692
(2,3-Dimethoxyphenyl)acetonitrile ≥98%
(2,3-Dimethoxyphenyl)acetonitrile
Hazard code(s): Irritant-Harmful


Formula: C₁₀H₁₁NO₂
Boiling Pt: 125 °C (1mm)
Density: 1.13 g/cm³
MDL Number: MFCD00016387
CAS Number: 4468-57-9
Order Now
Specification Test Results
Boiling point 125°C/1mm
Density 1.13

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty