Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Bromoquinoline

Supplier: MATRIX SCIENTIFIC MS
Warning

004184-5G 004184-25G
101786-904EA 29.23 USD InStock
101786-904 101786-906
8-Bromoquinoline
8-Bromoquinoline
Hazard code(s): Light Sensitive, Air Sensitive, Refrigerate, Stored under Argon


Formula: C₉H₆BrN
Boiling Pt: 302…304 °C
Density: 1.594 g/cm³
MDL Number: MFCD00191859
CAS Number: 16567-18-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty