Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromoanisole ≥98%

Supplier: MATRIX SCIENTIFIC MS

004235-100G 004235-500G
101787-004EA 100 USD InStock
101787-004 101787-006
2-Bromoanisole ≥98%
2-Bromoanisole
Hazard code(s): Irritant


Formula: C₇H₇BrO
Boiling Pt: 223 °C (1013 hPa)
Melting Pt: 2 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000064
CAS Number: 578-57-4
EINECS: 209-425-8
UN: 3082
ADR: 9,III
Order Now
Specification Test Results
Boiling point 223°C
Melting point 2°C
Density 1.502
ND20 1.574

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty