Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3',4'-Difluoroacetophenone ≥97%

Supplier: MATRIX SCIENTIFIC MS

004390-25G 004390-100G
101787-156EA 21.54 USD InStock
101787-156 101787-158
3',4'-Difluoroacetophenone ≥97%
3',4'-Difluoroacetophenone
Hazard code(s): Irritant


Formula: C₈H₆F₂O
Boiling Pt: 207 °C (1013 hPa)
Melting Pt: 20 °C
Density: 1.24 g/cm³ (20 °C)
MDL Number: MFCD00009891
CAS Number: 369-33-5
EINECS: 206-717-7
Order Now
Specification Test Results
Boiling point 94-95°C/13m
Melting point 19-20°C
Density 1.246
ND20 1.4916

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty