Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-4-fluorobenzoate ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

004395-25G 004395-100G
101787-174EA 21.54 USD InStock
101787-174 101787-176
Ethyl-4-fluorobenzoate ≥98% for synthesis
Ethyl-4-fluorobenzoate
Hazard code(s): Irritant


Formula: C₉H₉FO₂
Boiling Pt: 210 °C
Melting Pt: 25…27 °C
Density: 1.146 g/cm³ (20 °C)
MDL Number: MFCD00000351
CAS Number: 451-46-7
EINECS: 207-194-8
Merck Index: 13,04198
Order Now
Specification Test Results
Boiling point 210°C
Melting point 25-27°C
Density 1.146
ND20 1.486

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty