Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-(Trifluoromethyl)phenyl)imidazole ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

004412-1G 004412-5G
101787-200EA 207.69 USD InStock
101787-200 101787-202
1-(2-(Trifluoromethyl)phenyl)imidazole ≥98% for synthesis
1-(2-(Trifluoromethyl)phenyl)imidazole
Hazard code(s): Irritant, Light Sensitive


Formula: C₁₀H₇F₃N₂
Boiling Pt: 75 °C (0.25mm)
Melting Pt: 51…53 °C
MDL Number: MFCD00041206
CAS Number: 25371-96-4
Order Now
Specification Test Results
Boiling point 75°C/0.25mm
Melting point 51-53°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty